Assay Details
| Gene Symbol: MYB85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248877.1 | NP_001235806.1 | 2 - 3 | 375 | 74 | |
| GenBank mRNA | DQ822899.1 | - | 2 - 3 | 375 | 74 | |
More Information
Set Membership: |
|

MYB85
Genomic Map