Assay Details
| Gene Symbol: MYB85 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248877.1 | NP_001235806.1 | 4 - 5 | 669 | 87 | |
| GenBank mRNA | DQ822899.1 | - | 4 - 5 | 669 | 87 | |
More Information
Set Membership: |
|

MYB85
Genomic Map