Assay Details
| Gene Symbol: MYB107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250815.1 | NP_001237744.1 | 5 - 6 | 860 | 105 | |
| GenBank mRNA | DQ822909.1 | - | 5 - 6 | 860 | 105 | |
More Information
Set Membership: |
|

MYB107
Genomic Map