Assay Details
| Gene Symbol: MYB112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248213.1 | NP_001235142.1 | 1 | 657 | 102 | |
| GenBank mRNA | DQ822911.1 | - | 1 | 657 | 102 | |
More Information
Set Membership: |
|

MYB112
Genomic Map