Assay Details
| Gene Symbol: MYB118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248980.1 | NP_001235909.1 | 3 - 4 | 465 | 81 | |
| GenBank mRNA | DQ822912.1 | - | 3 - 4 | 465 | 81 | |
More Information
Set Membership: |
|

MYB118
Genomic Map