Assay Details
| Gene Symbol: MYB136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249038.1 | NP_001235967.1 | 5 - 6 | 710 | 85 | |
| GenBank mRNA | DQ822917.1 | - | 5 - 6 | 710 | 85 | |
More Information
Set Membership: |
|

MYB136
Genomic Map