Assay Details
| Gene Symbol: MYB136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249038.1 | NP_001235967.1 | 6 - 7 | 822 | 97 | |
| GenBank mRNA | DQ822917.1 | - | 6 - 7 | 822 | 97 | |
| Gene Symbol: LOC100783949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253119.2 | NP_001240048.1 | 6 - 7 | 97 | ||
| GenBank mRNA | BT093234.1 | - | 6 - 7 | 97 | ||
More Information
Set Membership: |
|

MYB136
Genomic Map