Assay Details
| Gene Symbol: MYB177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249137.1 | NP_001236066.1 | 5 - 6 | 754 | 93 | |
| GenBank mRNA | AK243995.1 | - | 5 - 6 | 93 | ||
| AK244532.1 | - | 5 - 6 | 93 | |||
| DQ822925.1 | - | 5 - 6 | 754 | 93 | ||
More Information
Set Membership: |
|

MYB177
Genomic Map