Assay Details
| Gene Symbol: MYB73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250849.1 | NP_001237778.1 | 2 - 3 | 192 | 86 | |
| GenBank mRNA | BT091291.1 | - | 2 - 3 | 216 | 86 | |
| DQ822927.1 | - | 2 - 3 | 192 | 86 | ||
More Information
Set Membership: |
|

MYB73
Genomic Map