Assay Details
| Gene Symbol: HIDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250299.2 | NP_001237228.1 | 1 | 64 | 88 | |
| GenBank mRNA | AB154415.1 | - | 1 | 55 | 88 | |
| AK245894.1 | - | 1 | 65 | 88 | ||
| BT097440.1 | - | 1 | 66 | 88 | ||
More Information
Set Membership: |
|

HIDH
Genomic Map