Assay Details
| Gene Symbol: LOC732556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249985.2 | NP_001236914.1 | 1 - 2 | 1226 | 74 | |
| GenBank mRNA | AB197676.1 | - | 1 - 2 | 1226 | 74 | |
| AK244653.1 | - | 1 - 2 | 1233 | 74 | ||
More Information
Set Membership: |
|

LOC732556
Genomic Map