Assay Details
Gene Symbol: BMY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249435.1 | NP_001236364.1 | 2 - 3 | 869 | 67 | |
GenBank mRNA | AJ871580.1 | - | 2 - 3 | 869 | 67 | |
AK244990.1 | - | 2 - 3 | 884 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
