Assay Details
| Gene Symbol: MSH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251288.1 | NP_001238217.1 | 18 - 19 | 2222 | 90 | |
| GenBank mRNA | AY856369.1 | - | 18 - 19 | 2222 | 90 | |
More Information
Set Membership: |
|

MSH1
Genomic Map