Assay Details
| Gene Symbol: LOC732542 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249779.1 | NP_001236708.1 | 8 - 9 | 1195 | 98 | |
| GenBank mRNA | AY954628.1 | - | 8 - 9 | 1143 | 98 | |
| AY954629.1 | - | 8 - 9 | 1195 | 98 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC732542
Genomic Map