Assay Details
| Gene Symbol: NAC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249942.2 | NP_001236871.1 | 1 - 2 | 216 | 93 | |
| GenBank mRNA | AK244823.1 | - | 1 - 2 | 228 | 93 | |
| DQ028770.1 | - | 1 - 2 | 218 | 93 | ||
| JX569048.1 | - | 1 - 2 | 157 | 93 | ||
More Information
Set Membership: |
|

NAC2
Genomic Map