Assay Details
| Gene Symbol: MT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251383.1 | NP_001238312.1 | 3 - 4 | 394 | 96 | |
| GenBank mRNA | AK244797.1 | - | 3 - 4 | 439 | 96 | |
| DQ278438.1 | - | 3 - 4 | 394 | 96 | ||
More Information
Set Membership: |
|

MT2
Genomic Map