Assay Details
| Gene Symbol: EN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248095.1 | NP_001235024.1 | 5 - 6 | 537 | 100 | |
| GenBank mRNA | EF521873.1 | - | 5 - 6 | 537 | 100 | |
More Information
Set Membership: |
|

EN2
Genomic Map