Assay Details
| Gene Symbol: OAS-TL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248188.1 | NP_001235117.1 | 4 - 5 | 729 | 73 | |
| GenBank mRNA | EF584898.1 | - | 4 - 5 | 729 | 73 | |
More Information
Set Membership: |
|

OAS-TL3
Genomic Map