Assay Details
| Gene Symbol: CCT-D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250475.1 | NP_001237404.1 | 1 | 1551 | 65 | |
| GenBank mRNA | AB004233.1 | - | 1 | 1551 | 65 | |
| AK286883.1 | - | 1 | 1621 | 65 | ||
More Information
Set Membership: |
|

CCT-D
Genomic Map