Assay Details
| Gene Symbol: PETC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250719.1 | NP_001237648.1 | 2 - 3 | 325 | 91 | |
| GenBank mRNA | AM498291.1 | - | 2 - 3 | 325 | 91 | |
| Gene Symbol: LOC100799420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289339.1 | NP_001276268.1 | 2 - 3 | 290 | 91 | |
| GenBank mRNA | AK243821.1 | - | 2 - 3 | 387 | 91 | |
| AK244194.1 | - | 2 - 3 | 316 | 91 | ||
| AK286526.1 | - | 2 - 3 | 390 | 91 | ||
| BT096467.1 | - | 2 - 3 | 290 | 91 | ||
More Information
Set Membership: |
|

PETC
Genomic Map