Assay Details
| Gene Symbol: TOC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248273.1 | NP_001235202.1 | 2 - 3 | 328 | 95 | |
| GenBank mRNA | EU076435.1 | - | 2 - 3 | 328 | 95 | |
More Information
Set Membership: |
|

TOC1
Genomic Map