Assay Details
| Gene Symbol: LOC100127416 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250666.1 | NP_001237595.1 | 4 - 5 | 1131 | 95 | |
| GenBank mRNA | EU169384.1 | - | 4 - 5 | 1131 | 95 | |
More Information
Set Membership: |
|

LOC100127416
Genomic Map