Assay Details
| Gene Symbol: LOC100127418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250701.1 | NP_001237630.1 | 4 - 5 | 729 | 102 | |
| GenBank mRNA | EU169386.1 | - | 4 - 5 | 729 | 102 | |
| M93140.1 | - | 5 - 6 | 809 | 102 | ||
More Information
Set Membership: |
|

LOC100127418
Genomic Map