Assay Details
| Gene Symbol: LOC100127418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250701.1 | NP_001237630.1 | 5 - 6 | 810 | 87 | |
| GenBank mRNA | EU169386.1 | - | 5 - 6 | 810 | 87 | |
| M93140.1 | - | 6 - 7 | 890 | 87 | ||
More Information
Set Membership: |
|

LOC100127418
Genomic Map