Assay Details
| Gene Symbol: LOC100170725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251402.1 | NP_001238331.1 | 3 - 4 | 740 | 87 | |
| GenBank mRNA | AB331958.1 | - | 3 - 4 | 740 | 87 | |
More Information
Set Membership: |
|

LOC100170725
Genomic Map