Assay Details
| Gene Symbol: LOC100170728 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251448.1 | NP_001238377.1 | 3 - 4 | 228 | 98 | |
| GenBank mRNA | AB331962.1 | - | 3 - 4 | 228 | 98 | |
| AK287393.1 | - | 3 - 4 | 310 | 98 | ||
| BT095890.1 | - | 3 - 4 | 302 | 98 | ||
| Gene Symbol: LOC100777928 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287282.1 | - | 3 - 4 | 307 | 98 | |
More Information
Set Membership: |
|

LOC100170728
Genomic Map