Assay Details
Gene Symbol: PHR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001251781.2 | NP_001238710.1 | 1 - 2 | 320 | 106 | |
GenBank mRNA | AB359233.1 | - | 1 - 2 | 371 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
