Assay Details
| Gene Symbol: PHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251781.2 | NP_001238710.1 | 2 - 3 | 668 | 66 | |
| GenBank mRNA | AB359233.1 | - | 2 - 3 | 719 | 66 | |
More Information
Set Membership: |
|

PHR
Genomic Map