Assay Details
| Gene Symbol: LOC100170690 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250822.1 | NP_001237751.1 | 1 | 251 | 91 | |
| GenBank mRNA | BT089152.1 | - | 1 | 271 | 91 | |
| EU444601.1 | - | 1 | 251 | 91 | ||
More Information
Set Membership: |
|

LOC100170690
Genomic Map