Assay Details
| Gene Symbol: NAC22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289358.1 | NP_001276287.1 | 1 - 2 | 201 | 70 | |
| GenBank mRNA | EU661921.1 | - | 1 - 2 | 398 | 70 | |
| JX569045.1 | - | 1 - 2 | 201 | 70 | ||
More Information
Set Membership: |
|

NAC22
Genomic Map