Assay Details
Gene Symbol: LOC100194420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249643.1 | NP_001236572.1 | 1 - 2 | 359 | 89 | |
GenBank mRNA | AK286805.1 | - | 1 - 2 | 409 | 89 | |
BT097708.1 | - | 1 - 2 | 415 | 89 | ||
FJ222387.1 | - | 1 - 2 | 359 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
