Assay Details
Gene Symbol: LOC100194420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249643.1 | NP_001236572.1 | 4 - 5 | 656 | 106 | |
GenBank mRNA | AK286805.1 | - | 4 - 5 | 709 | 106 | |
BT097708.1 | - | 4 - 5 | 715 | 106 | ||
FJ222387.1 | - | 4 - 5 | 656 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
