Assay Details
Gene Symbol: LOC100194420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001249643.1 | NP_001236572.1 | 7 - 8 | 980 | 86 | |
GenBank mRNA | AK286805.1 | - | 7 - 8 | 1033 | 86 | |
BT097708.1 | - | 7 - 8 | 1039 | 86 | ||
FJ222387.1 | - | 7 - 8 | 980 | 86 |
Gene Symbol: LOC100784022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK243740.1 | - | 7 - 8 | 1176 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
