Assay Details
| Gene Symbol: ICE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251648.1 | NP_001238577.1 | 3 - 4 | 1197 | 98 | |
| GenBank mRNA | FJ393224.1 | - | 3 - 4 | 1197 | 98 | |
More Information
Set Membership: |
|

ICE2
Genomic Map