Assay Details
| Gene Symbol: LOC548057 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251210.1 | NP_001238139.1 | 1 | 154 | 88 | |
| GenBank mRNA | AY439102.1 | - | 1 | 154 | 88 | |
| Gene Symbol: LOC100306118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250898.1 | NP_001237827.1 | 1 | 267 | 88 | |
| GenBank mRNA | BT090069.1 | - | 1 | 267 | 88 | |
More Information
Set Membership: |
|

LOC548057
Genomic Map