Assay Details
| Gene Symbol: MYB62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248772.1 | NP_001235701.1 | 1 | 251 | 112 | |
| GenBank mRNA | DQ822889.1 | - | 1 | 251 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYB62| Gene Symbol: MYB62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248772.1 | NP_001235701.1 | 1 | 251 | 112 | |
| GenBank mRNA | DQ822889.1 | - | 1 | 251 | 112 | |
Set Membership: |
|