Assay Details
| Gene Symbol: HSP90-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001249683.1 | NP_001236612.1 | 3 | 2044 | 84 | |
| GenBank mRNA | FJ222390.1 | - | 3 | 2044 | 84 | |
| FJ969430.1 | - | 3 | 2044 | 84 | ||
More Information
Set Membership: |
|

HSP90-1
Genomic Map