Assay Details
| Gene Symbol: SCAM-2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250954.1 | NP_001237883.1 | 2 | 368 | 102 | |
| GenBank mRNA | BT095754.1 | - | 2 | 368 | 102 | |
| L01431.1 | - | 2 | 349 | 102 | ||
More Information
Set Membership: |
|

SCAM-2
Genomic Map