Assay Details
| Gene Symbol: PRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251664.1 | NP_001238593.1 | 1 | 650 | 107 | |
| GenBank mRNA | BT092649.1 | - | 1 | 650 | 107 | |
| S44202.1 | - | 1 | 654 | 107 | ||
More Information
Set Membership: |
|

PRP2
Genomic Map