Assay Details
| Gene Symbol: PRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250441.2 | NP_001237370.1 | 1 | 127 | 64 | |
| GenBank mRNA | BT090793.1 | - | 1 | 129 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRP3| Gene Symbol: PRP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250441.2 | NP_001237370.1 | 1 | 127 | 64 | |
| GenBank mRNA | BT090793.1 | - | 1 | 129 | 64 | |
Set Membership: |
|