Assay Details
| Gene Symbol: FAD7-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001250909.2 | NP_001237838.2 | 4 - 5 | 1343 | 81 | |
| GenBank mRNA | AK244078.1 | - | 4 - 5 | 1343 | 81 | |
| GQ144962.1 | - | 4 - 5 | 1190 | 81 | ||
More Information
Set Membership: |
|

FAD7-1
Genomic Map