Assay Details
| Gene Symbol: CYN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001248706.2 | NP_001235635.1 | 1 - 2 | 296 | 103 | |
| GenBank mRNA | AK245550.1 | - | 1 - 2 | 279 | 103 | |
| BT089835.1 | - | 1 - 2 | 300 | 103 | ||
More Information
Set Membership: |
|

CYN
Genomic Map