Assay Details
| Gene Symbol: LOC100792337 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252792.2 | NP_001239721.1 | 1 - 2 | 198 | 59 | |
| GenBank mRNA | BT098672.1 | - | 5 - 6 | 582 | 59 | |
More Information
Set Membership: |
|

LOC100792337
Genomic Map