Assay Details
| Gene Symbol: LOC100813383 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253085.2 | NP_001240014.1 | 1 - 2 | 195 | 104 | |
| GenBank mRNA | BT098151.1 | - | 5 - 6 | 806 | 104 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001253085.2 transcripts.
Set Membership: |
|

LOC100813383
Genomic Map