Assay Details
| Gene Symbol: LOC100808776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253091.2 | NP_001240020.1 | 4 - 5 | 330 | 86 | |
| GenBank mRNA | BT095106.1 | - | 5 - 6 | 684 | 86 | |
| BT097621.1 | - | 4 - 5 | 332 | 86 | ||
More Information
Set Membership: |
|

LOC100808776
Genomic Map