Assay Details
| Gene Symbol: LOC100804707 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253234.2 | NP_001240163.1 | 5 - 6 | 661 | 65 | |
| GenBank mRNA | BT094050.1 | - | 5 - 6 | 663 | 65 | |
More Information
Set Membership: |
|

LOC100804707
Genomic Map