Assay Details
| Gene Symbol: LOC100788071 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253261.1 | NP_001240190.1 | 1 - 2 | 285 | 101 | |
| GenBank mRNA | AK286990.1 | - | 1 - 2 | 267 | 101 | |
| BT098288.1 | - | 1 - 2 | 285 | 101 | ||
More Information
Set Membership: |
|

LOC100788071
Genomic Map