Assay Details
| Gene Symbol: LOC100807528 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253263.2 | NP_001240192.1 | 3 - 4 | 546 | 80 | |
| GenBank mRNA | BT094052.1 | - | 3 - 4 | 548 | 80 | |
| FJ594401.1 | - | 3 - 4 | 501 | 80 | ||
More Information
Set Membership: |
|

LOC100807528
Genomic Map