Assay Details
| Gene Symbol: LOC100799090 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253311.1 | NP_001240240.1 | 6 - 7 | 776 | 75 | |
| GenBank mRNA | BT097142.1 | - | 6 - 7 | 776 | 75 | |
More Information
Set Membership: |
|

LOC100799090
Genomic Map