Assay Details
| Gene Symbol: LOC100775298 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253990.1 | NP_001240919.1 | 4 - 5 | 347 | 64 | |
| GenBank mRNA | BT096425.1 | - | 4 - 5 | 347 | 64 | |
More Information
Set Membership: |
|

LOC100775298
Genomic Map